DL-Serine Hydrazide Hydrochloride 97%

Supplier: Ambeed
Warning

A470145-100G A470145-1G A470145-25G A470145-5G
76979-658EA 149 USD
76979-658 76979-660 76979-662 76979-664
DL-Serine Hydrazide Hydrochloride 97%
DL-Serine Hydrazide Hydrochloride

Precaución: for Research Use Only

Formula: C₃H₁₀ClN₃O₂
MW: 155.58 g/mol
MDL Number: MFCD08704793
CAS Number: 55819-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR