2-Chloro-4-(methoxymethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A131508-100MG A131508-1G A131508-250MG
76972-096EA 106.09 USD
76972-096 76972-098 76972-100
2-Chloro-4-(methoxymethyl)pyrimidine 97%
2-Chloro-4-(methoxymethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.58 g/mol
MDL Number: MFCD18837162
CAS Number: 1289387-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR