Z-Asp(OtBu)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-Asp(OtBu)-OH, N-Benzyloxycarbonyl-L-aspartic acid 4-tert-butyl ester

A339928-100G A339928-1G A339928-25G A339928-5G
76978-378EA 173.99 USD
76978-378 76978-380 76978-382 76978-384
Z-Asp(OtBu)-OH 98%
Z-Asp(OtBu)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
MDL Number: MFCD00042857
CAS Number: 5545-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR