2,6-Diaminopyrazine 98+%

Supplier: Ambeed
Warning

A520122-100MG A520122-1G A520122-250MG
76980-004EA 38.2 USD
76980-004 76980-006 76980-008
2,6-Diaminopyrazine 98+%
2,6-Diaminopyrazine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MDL Number: MFCD09907864
CAS Number: 41536-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR