(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2S,5S)-5-Benzyl-2-(tert-butyl)-3-methylimidazolidin-4-one, (2S,5S)-(-)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone

A197618-1G A197618-250MG A197618-5G A197618-100MG
76975-116EA 175.05 USD
76975-116 76975-118 76975-120 77671-750
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97%, ee 99%
(2S,5S)-(-)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O
MW: 246.35 g/mol
MDL Number: MFCD03426982
CAS Number: 346440-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR