(-)-2,6-Bis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazolin-2-yl]pyridine ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Bis((3aS,8aR)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)pyridine, Indenyl-PYBOX
A132781-100MG
A132781-1G
A132781-250MG
A132781-25G
A132781-5G
76972-170EA
7
USD
76972-170
76972-172
76972-174
76972-176
76972-178
(-)-2,6-Bis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazolin-2-yl]pyridine ≥98%, ee 99%
(-)-2,6-Bis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazolin-2-yl]pyridine
Precaución: for Research Use Only
Formula:
C₂₅H₁₉N₃O₂ |
MDL Number:
MFCD05861599 CAS Number: 185346-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...