2,6-Bis((S)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%

Supplier: Ambeed
Warning

A442966-100MG A442966-1G A442966-250MG
76979-384EA 145 USD
76979-384 76979-386 76979-388
2,6-Bis((S)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine ≥98%, ee 99%
2,6-Bis((S)-4-isopropyl-5,5-diphenyl-4,5-dihydrooxazol-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₄₁H₃₉N₃O₂
MDL Number: MFCD18379147
CAS Number: 162213-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR