2-Chloro-5-methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A215065-100MG A215065-1G A215065-250MG
76975-648EA 49.87 USD
76975-648 76975-650 76975-652
2-Chloro-5-methylbenzo[d]thiazole 95%
2-Chloro-5-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MW: 183.65 g/mol
MDL Number: MFCD09260926
CAS Number: 3622-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR