6-Aminopenicillanic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-APA 6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-amino-2,2-dimethyl-5-oxoperhydroazeto[2,1-b][1,3]thiazole-3-carboxylic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (+)-6-Aminopenicillanic acid, (2S,5R,6R)-(+)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid

A141395-500G A141395-100G A141395-1000G
76972-622EA 81.69 USD
76972-622 77611-290 77671-018
6-Aminopenicillanic acid 95%
6-Aminopenicillanic acid

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₃S
MW: 216.26 g/mol
MDL Number: MFCD00005176
CAS Number: 551-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR