Acitretin 98%

Supplier: Ambeed
Danger

A470794-100MG A470794-250MG A470794-50MG A470794-10MG A470794-1MG A470794-25G A470794-5G
76979-666EA 15 USD
76979-666 76979-668 76979-670 77613-456 77673-210 77673-212 77673-214
Acitretin 98%
Acitretin

Precaución: for Research Use Only

Formula: C₂₁H₂₆O₃
MDL Number: MFCD00866632
CAS Number: 55079-83-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR