(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

A238915-10G A238915-1G A238915-250MG A238915-5G
76976-388EA 73 USD
76976-388 76976-390 76976-392 77881-626
(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride 97%
(S)-3-Amino-1-carbobenzoxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD06796559
CAS Number: 550378-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR