5-Chloro-2-methoxy-nicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-methoxypyridine-3-carboxylic acid, 5-Chloro-2-methoxynicotinic acid

A158214-10G A158214-1G A158214-25G A158214-5G
76973-414EA 55 USD
76973-414 76973-416 76973-418 76973-420
5-Chloro-2-methoxy-nicotinic acid 98%
5-Chloro-2-methoxy-nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD08236790
CAS Number: 54916-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR