4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(5-Methyl-1,3,4-oxadiazol-2-yl)phenyl)boronic acid

A364945-100MG A364945-1G A364945-250MG
76978-638EA 47 USD
76978-638 76978-640 76978-642
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 98%
4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₃
MDL Number: MFCD08436062
CAS Number: 913835-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR