Dasatinib monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dasatinib hydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide monohydrate, N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrate
A210354-1MG
A210354-100MG
A210354-10G
A210354-50MG
A210354-5G
76975-514EA
5.31
USD
76975-514
77671-866
77671-868
77671-870
77671-872
Dasatinib monohydrate 98%
Dasatinib monohydrate
Precaución: for Research Use Only
Formula:
C₂₂H₂₆ClN₇O₂S·H₂O MW: 506.02 g/mol |
MDL Number:
MFCD08704581 CAS Number: 863127-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...