Acetohydroxamic acid 98%

Supplier: Ambeed
Danger

A168949-100MG A168949-10G A168949-25G A168949-5G A168949-1000G A168949-100G A168949-500G
76973-894EA 5 USD
76973-894 76973-896 76973-898 76973-900 77550-088 77593-040 77671-384
Acetohydroxamic acid 98%
Acetohydroxamic acid

Precaución: for Research Use Only

Formula: C₂H₅NO₂
MDL Number: MFCD00009994
CAS Number: 546-88-3
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR