tert-Butyl (1-(6-bromopyridin-2-yl)piperidin-4-yl)carbamate 97%

Supplier: Ambeed
Warning

A277594-1G A277594-250MG
76977-322EA 240 USD
76977-322 76977-324
tert-Butyl (1-(6-bromopyridin-2-yl)piperidin-4-yl)carbamate 97%
tert-Butyl (1-(6-bromopyridin-2-yl)piperidin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₂BrN₃O₂
MDL Number: MFCD10698891
CAS Number: 848500-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR