Ethyl-2-chloroisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-Chloropyridine-4-carboxylate

A127212-100G A127212-10G A127212-25G A127212-5G
76971-922EA 117.76 USD
76971-922 76971-924 76971-926 76971-928
Ethyl-2-chloroisonicotinate 98%
Ethyl 2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD04114259
CAS Number: 54453-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR