3-Bromo-2-nitropyridine 98%

Supplier: Ambeed
Warning

A139378-100G A139378-10G A139378-1G A139378-250MG A139378-5G
76972-518EA 570.77 USD
76972-518 76972-520 76972-522 76972-524 77670-992
3-Bromo-2-nitropyridine 98%
3-Bromo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD00955614
CAS Number: 54231-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR