2-Bromo-1-(1H-pyrrol-2-yl)ethanone 95%

Supplier: Ambeed
Danger

A186104-100MG A186104-250MG A186104-1G
76974-632EA 54.11 USD
76974-632 76974-634 77671-628
2-Bromo-1-(1H-pyrrol-2-yl)ethanone 95%
2-Bromo-1-(1H-pyrrol-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD11182402
CAS Number: 73742-16-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR