2-Chloro-5-(trifluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A142164-100MG A142164-10G A142164-1G A142164-5G
76972-650EA 48.81 USD
76972-650 76972-652 76972-654 76972-656
2-Chloro-5-(trifluoromethoxy)pyridine 95%
2-Chloro-5-(trifluoromethoxy)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃NO
MW: 197.542 g/mol
MDL Number: MFCD18257007
CAS Number: 1206972-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR