2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine 97%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2-(5-Fluoro-2-oxo-4-(((pentyloxy)carbonyl)amino)pyrimidin-1(2H)-yl)-5-methyltetrahydrofuran-3,4-diyl diacetate, 5'-Deoxy-5-fluoro-N-[(pentyloxy)carbonyl]cytidine 2',3'-Diacetate

A253874-10G A253874-250MG A253874-25G A253874-5G
76976-734EA 31.83 USD
76976-734 76976-736 76976-738 76976-740
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine 97%
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine

Precaución: for Research Use Only

Formula: C₁₉H₂₆FN₃O₈
MW: 443.43 g/mol
MDL Number: MFCD07369303
CAS Number: 162204-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR