Sodium 2,5-difluorobenzoate 95+%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluorobenzoic acid sodium salt

A495565-100G A495565-25G A495565-5G
76979-810EA 178.24 USD
76979-810 76979-812 76979-814
Sodium 2,5-difluorobenzoate 95+%
Sodium 2,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₇H₃F₂NaO₂
MW: 180.08 g/mol
MDL Number: MFCD03094396
CAS Number: 522651-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR