6-Bromo-3-fluoroisoquinoline 95%

Supplier: Ambeed
Warning

A181046-100MG A181046-1G A181046-250MG
76974-458EA 29 USD
76974-458 77593-154 77881-372
6-Bromo-3-fluoroisoquinoline 95%
6-Bromo-3-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MDL Number: MFCD21603999
CAS Number: 891785-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR