6-Chloro-1,3-dihydro-2H-pyrrolo[3,2-c]pyridin-2-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-1H-pyrrolo[3,2-c]pyridin-2(3H)-one, 6-Chloro-2,3-dihydro-2-oxo-1H-pyrrolo[3,2-c]pyridine
A140559-100MG
A140559-1G
A140559-250MG
A140559-5G
76972-580EA
63.66
USD
76972-580
76972-582
76972-584
77671-004
6-Chloro-1,3-dihydro-2H-pyrrolo[3,2-c]pyridin-2-one 98%
6-Chloro-1,3-dihydro-2H-pyrrolo[3,2-c]pyridin-2-one
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂O MW: 168.58 g/mol |
MDL Number:
MFCD08690138 CAS Number: 1000342-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...