7-Bromo-5-chloro-4-azaindole 95%

Supplier: Ambeed
Warning

A360227-100MG A360227-1G A360227-250MG A360227-50MG A360227-5G
76978-580EA 143 USD
76978-580 77551-030 77672-808 77881-980 77881-981
7-Bromo-5-chloro-4-azaindole 95%
7-Bromo-5-chloro-4-azaindole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MDL Number: MFCD11845520
CAS Number: 1082040-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR