1-Methyl-1H-pyrrole-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A124048-100MG A124048-250MG A124048-1G
76971-724EA 44.56 USD
76971-724 76971-726 77566-372
1-Methyl-1H-pyrrole-3-carbaldehyde 95%
1-Methyl-1H-pyrrole-3-carbaldehyde

Precaución: For research use only.

Formula: C₆H₇NO
MW: 109.12 g/mol
MDL Number: MFCD02661657
CAS Number: 36929-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR