(6R,7R)-7-Amino-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Amino-3-chloro cephalosporanic acid
A117658-100MG
A117658-10G
A117658-1G
A117658-250MG
A117658-5G
76971-304EA
20.16
USD
76971-304
76971-306
76971-308
76971-310
76971-312
(6R,7R)-7-Amino-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid 96%
(6R,7R)-7-Amino-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₇ClN₂O₃S MW: 234.66 g/mol |
MDL Number:
MFCD09039101 CAS Number: 53994-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...