5-(Bromomethyl)-2-methylpyrimidin-4-amine hydrobromide 95%

Supplier: Ambeed
Danger

A250353-100MG A250353-1G A250353-250MG A250353-5G
76976-672EA 201.58 USD
76976-672 76976-674 76976-676 76976-678
5-(Bromomethyl)-2-methylpyrimidin-4-amine hydrobromide 95%
5-(Bromomethyl)-2-methylpyrimidin-4-amine hydrobromide

Precaución: for Research Use Only

Formula: C₆H₉Br₂N₃
MW: 282.96 g/mol
MDL Number: MFCD05149311
CAS Number: 2908-71-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR