Ethyl-4,6-dichloropyrimidine-5-carboxylate 95%

Supplier: Ambeed
Danger

A772288-100MG A772288-1G A772288-5G
76981-516EA 40 USD
76981-516 76981-518 76981-520
Ethyl-4,6-dichloropyrimidine-5-carboxylate 95%
Ethyl-4,6-dichloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MDL Number: MFCD10698803
CAS Number: 87600-72-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR