4-Fluoro-3-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A543929-100MG A543929-1G A543929-250MG
76980-196EA 21.22 USD
76980-196 76980-198 76980-200
4-Fluoro-3-hydroxybenzaldehyde 98%
4-Fluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅FO₂
MW: 140.112 g/mol
MDL Number: MFCD11110190
CAS Number: 103438-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR