3-Iodopyrazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A494014-1G A494014-250MG
76979-800EA 27.59 USD
76979-800 76979-802 76979-804
3-Iodopyrazine-2-carboxylic acid 95%
3-Iodopyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C5H3IN2O2
MDL Number: MFCD13193281
CAS Number: 212471-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR