Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Asn(Trt)-OH, Nɑ-Boc-Nγ-trityl-L-asparagine

A164982-100G A164982-10G A164982-1G A164982-25G A164982-5G
76973-742EA 156 USD
76973-742 76973-744 76973-746 76973-748 76973-750
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine 97%
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₂O₅
MDL Number: MFCD00153299
CAS Number: 132388-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR