3,3-Dimethylpyrrolidine hydrochloride 98%

Supplier: Ambeed
Warning

A859042-250MG A859042-1G A859042-100MG
76981-826EA 55 USD
76981-826 77614-538 77882-783
3,3-Dimethylpyrrolidine hydrochloride 98%
3,3-Dimethylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃N·ClH
MDL Number: MFCD11849606
CAS Number: 3437-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR