2-Amino-6-chloronicotinic acid 95%

Supplier: Ambeed
Warning

A147450-10G A147450-1G A147450-250MG A147450-5G A147450-25G
76972-894EA 188.85 USD
76972-894 76972-896 76972-898 76972-900 77671-078
2-Amino-6-chloronicotinic acid 95%
2-Amino-6-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.5700 g/mol
MDL Number: MFCD11045328
CAS Number: 58584-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR