2-(Trifluoromethyl)-p-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-2-(trifluoromethyl)aniline

A153101-10G A153101-1G A153101-5G
76973-188EA 48.81 USD
76973-188 76973-190 76973-192
2-(Trifluoromethyl)-p-anisidine 98%
2-(Trifluoromethyl)-p-anisidine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD04972660
CAS Number: 53903-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR