1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl 3-ethyl 4-aminopiperidine-1,3-dicarboxylate

A705857-100MG A705857-250MG
76981-136EA 112.46 USD
76981-136 76981-138
1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate 95%
1-tert-Butyl-3-ethyl-4-aminopiperidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₄
MW: 272.3407 g/mol
MDL Number: MFCD11877860
CAS Number: 932035-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR