N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide 95%

Supplier: Ambeed
Warning

A320343-1G A320343-5G
76978-066EA 26.53 USD
76978-066 76978-068
N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide 95%
N-(4-Bromo-1-methyl-1H-pyrazol-5-yl)pivalamide

Precaución: for Research Use Only

Formula: C₉H₁₄BrN₃O
MW: 260.1300 g/mol
MDL Number: MFCD01763678
CAS Number: 679394-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR