6-Aminoisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A277032-100MG A277032-1G A277032-250MG
76977-302EA 21 USD
76977-302 77672-424 77881-754
6-Aminoisoquinolin-1(2H)-one 97%
6-Aminoisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MDL Number: MFCD12547311
CAS Number: 216099-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR