4-Hydroxypiperidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Piperidinol Hydrochloride, Piperidin-4-ol hydrochloride

A355862-1G A355862-5G A355862-25G
76978-552EA 8 USD
76978-552 76978-554 77672-780
4-Hydroxypiperidine hydrochloride 97%
4-Hydroxypiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MDL Number: MFCD00035298
CAS Number: 5382-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR