8-Hydroxyquinoline-N-Oxide 97%

Supplier: Ambeed
Warning

A422679-100G A422679-10G A422679-1G A422679-25G A422679-5G
76979-242EA 127 USD
76979-242 76979-244 76979-246 76979-248 76979-250
8-Hydroxyquinoline-N-Oxide 97%
8-Hydroxyquinoline-N-Oxide

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD00006739
CAS Number: 1127-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR