4-Bromo-2,3-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A153150-1G A153150-250MG A153150-5G A153150-100MG A153150-25G
76973-198EA 27 USD
76973-198 76973-200 76973-202 77881-180 77881-181
4-Bromo-2,3-difluorobenzonitrile 97%
4-Bromo-2,3-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MDL Number: MFCD09030640
CAS Number: 126163-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR