tert-Butyl 3-(piperidin-4-yl)azetidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A195551-250MG A195551-50MG A195551-100MG
76975-014EA 303.42 USD
76975-014 76975-016 77671-732
tert-Butyl 3-(piperidin-4-yl)azetidine-1-carboxylate 95%
tert-Butyl 3-(piperidin-4-yl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.34 g/mol
MDL Number: MFCD14581437
CAS Number: 1251006-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR