1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 96%

Supplier: Ambeed
Danger

Synonyms: 1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutylpyrazole-4-boronic acid, pinacol ester

A435489-1G A435489-250MG A435489-5G
76979-338EA 47 USD
76979-338 77882-136 77882-137
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester 96%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₃BN₂O₂
MDL Number: MFCD05663857
CAS Number: 827614-66-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR