6-Bromopyridazine-3-carbonitrile 95%

Supplier: Ambeed
Warning

A590935-1G A590935-250MG
76980-436EA 393.6 USD
76980-436 76980-438
6-Bromopyridazine-3-carbonitrile 95%
6-Bromopyridazine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 183.99 g/mol
MDL Number: MFCD10688600
CAS Number: 1027513-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR