3-Bromo-4-methylbenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-methyl-4-(trifluoromethyl)benzene

A207007-100G A207007-10G A207007-1G A207007-25G A207007-5G
76975-380EA 600.47 USD
76975-380 76975-382 76975-384 76975-386 76975-388
3-Bromo-4-methylbenzotrifluoride 98%
3-Bromo-4-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631584
CAS Number: 66417-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR