β-Chloro-4'-fluoropropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

A112584-100G A112584-25G
76970-928EA 108.22 USD
76970-928 76970-930
β-Chloro-4'-fluoropropiophenone 97%
β-Chloro-4'-fluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈ClFO
MW: 186.61 g/mol
MDL Number: MFCD00000991
CAS Number: 347-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR