Zosuquidar trihydrochloride 98%

Supplier: Ambeed
Warning

A272992-10MG A272992-1MG A272992-25MG A272992-5MG
76977-220EA 147.47 USD
76977-220 76977-222 76977-224 76977-226
Zosuquidar trihydrochloride 98%
Zosuquidar trihydrochloride

Precaución: for Research Use Only

Formula: C₃₂H₃₁FN₃O·3HCl
MDL Number: MFCD00942300
CAS Number: 167465-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR