1-bromo-4-iodonaphthalene 98%

Supplier: Ambeed
Warning

A179023-100G A179023-10G A179023-250MG A179023-25G A179023-5G A179023-1G
76974-360EA 275 USD
76974-360 76974-362 76974-364 76974-366 76974-368 77671-532
1-bromo-4-iodonaphthalene 98%
1-bromo-4-iodonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrI
MDL Number: MFCD01166242
CAS Number: 63279-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR