(3-(Naphthalen-1-yl)phenyl)boronic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(1-Naphthyl)phenylboronic acid, 3-(1-Naphthyl)benzeneboronic acid
A112222-10G
A112222-5G
76970-894EA
169
USD
76970-894
76970-896
(3-(Naphthalen-1-yl)phenyl)boronic acid 96%
(3-(Naphthalen-1-yl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₃BO₂ MW: 248.088 g/mol |
MDL Number:
MFCD11045054 CAS Number: 881913-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...