(4-Phenylnaphthalen-1-yl)boronic acid 97%

Supplier: Ambeed
Warning

A125944-10G A125944-1G A125944-250MG A125944-5G
76971-866EA 47 USD
76971-866 76971-868 76971-870 76971-872
(4-Phenylnaphthalen-1-yl)boronic acid 97%
(4-Phenylnaphthalen-1-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃BO₂
MDL Number: MFCD16294428
CAS Number: 372521-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR